missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Direct Scarlet B (Technical Grade), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Direct Scarlet B (Technical Grade) |
| CAS | 3530-19-6 |
| Molekylformel | C30H22N4Na2O8S2 |
| Kvantitet | 250 mg |
| Synonym | 8-[2-[4'-[2-(4-ethoxyphenyl)diazenyl][1,1'-biphenyl]-4-yl]diazenyl]-7-hydroxy-1,3-Naphthalenedisulfonic acid sodium salt, 8-[[4’-[(4-ethoxyphenyl)azo][1,1'-biphenyl]-4-yl]azo]-7-hydroxy-1,3-Naphthalenedisulfonic acid disodium salt, C.I. Direct Red 37, disodium salt (8CI), Airedale Scarlet BD, Amanil Fast Scarlet B, Atlantic Scarlet B, Azine Scarlet BX, Benzanil Scarlet B, Benzanol Brilliant Scarlet B, Benzo Scarlet B, Benzyl Scarlet BS, C.I. 22240, C.I. Direct Red 37, Calcomine Scarlet B, Chloramine Red B, Chrome Leather Scarlet BS, Diamine Scarlet BA-CF, Diaphtamine Fast Red B, Diazol Scarlet B, Diphenyl Red BS, Diphenyl Scarlet BS, Direct Fast Scarlet B, Direct Fast Scarlet BS, Direct Scarlet BS, Erie Scarlet B, Fenamin Scarlet B, Kayaku Direct Scarlet B, Kayaku Direct Scarlet B New, Mitsui Direct Scarlet BS, Nippon Scarlet B, Nitto Direct Scarlet B, Paper Scarlet BAP, Paramine Fast Scarlet B, Phenamine Scarlet B, Pontamine Scarlet B, Shikiso Direct Scarlet BX, Tertrodirect Ponceau B, Triazol Fast Scarlet B, Union Fast Scarlet B |
| InChI formel | InChI=1S/C30H24N4O8S2.2Na/c1-2-42-25-14-12-24(13-15-25)32-31-22-8-3-19(4-9-22)20-5-10-23(11-6-20)33-34-30-27(35)16-7-21-17-26(43(36,37)38)18-28(29(21)30)44(39,40)41;;/h3-18,35H,2H2,1H3,(H,36,37,38)(H,39,40,41);;/q;2*+1/p-2 |
| LEDER | [Na+].[Na+].CCOc1ccc(cc1)N=Nc2ccc(cc2)c3ccc(cc3)N=Nc4c(O)ccc5cc(cc(c45)S(=O)(=O)[O-])S(=O)(=O)[O-] |
| IUPAC-namn | disodium;8-[(E)-[4-[4-[(E)-(4-ethoxyphenyl)diazenyl]phenyl]phenyl]diazenyl]-7-hydroxynaphthalene-1,3-disulfonate |
| Molekylvikt (g/mol) | 676.63 |
| Formel vikt | 676.0674 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?