Läs mer
Disecoctyl Phthalate, TRC
Produktkod. 30012868
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012868 100 mg
30007106 500 mg
17944535 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012868 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD429513100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 131-15-7
Molekylformel C24H38O4
Molekylvikt (g/mol) 390.56
LEDER CCCCCCC(C)OC(=O)C1=CC=CC=C1C(=O)OC(C)CCCCCC
Specifikationer
Kemiskt namn eller material Disecoctyl Phthalate
Kvantitet 100 mg
InChI formel InChI=1S/C24H38O4/c1-5-7-9-11-15-19(3)27-23(25)21-17-13-14-18-22(21)24(26)28-20(4)16-12-10-8-6-2/h13-14,17-20H,5-12,15-16H2,1-4H3
Formel vikt 390.277