Läs mer
MedChemExpress Disitertide
Produktkod. 16492639
Encompass_Preferred
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
16492639 1 mg
1 options
Produktkod. 16492639 Leverantör MedChemExpress Leverantörsnummer HYP0118
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
MedChemExpress Disitertide (P144) is a peptidic transforming growth factor-beta 1 (TGF-β1) inhibitor specifically designed to block the interaction with its receptor. Disitertide (P144) is also a PI3K inhibitor and an apoptosis inducer.
Specifikationer
CAS 272105-42-7
Innehåll och lagring Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
För användning med (applikation) Cancer-Kinase/protease
Molekylvikt (g/mol) 1580.82 g/mol
Produkttyp Reference compound
Kvantitet 1 mg
Frakt skick Room temperature
Synonym P144