Läs mer
Disodium Citrate, TRC
Produktkod. 30370076
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30370076 1 g
1 options
Produktkod. 30370076 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4941631G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Disodium Citrate
CAS 144-33-2
Molekylformel C6H6Na2O7
Kvantitet 1 g
Synonym Alkacitron, Citralka, Disodium Citrate, Disodium Hydrogen Citrate, Disodium Monohydrogen Citrate, Monobasic Sodium Citrate
InChI formel InChI=1S/C6H8O7.2Na/c7-3(8)1-6(13,5(11)12)2-4(9)10;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;/q;2*+1/p-2
LEDER OC(C(CC(O[Na])=O)(CC(O[Na])=O)O)=O
Molekylvikt (g/mol) 236.09