Läs mer
Disodium (Ethoxyoxydophosphanyl)formate, Mikromol™
Produktkod. 30052671
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30052671 25 mg
1 options
Produktkod. 30052671 Leverantör Mikromol Leverantörsnummer MM3500.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Disodium (Ethoxyoxydophosphanyl)formate
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Disodium (ethoxyoxydophosphanyl)formate
CAS 55920-24-6
Analyt- eller komponentnamn Disodium (ethoxyoxydophosphanyl)formate
Frakt skick Room Temperature
Molekylformel C3 H5 O5 P . 2 Na
Certifikat och efterlevnad ISO 9001
Synonym Phosphinecarboxylic acid, 1-ethoxy-1-hydroxy-, 1-oxide, sodium salt (1:2), Phosphinecarboxylic acid, ethoxyhydroxy-, oxide, disodium salt (9CI), Disodium (Ethoxyoxydophosphanyl)formate, Foscarnet Sodium Hexahydrate Imp. B (EP), Foscarnet Imp. B (EP), Foscarnet Related Compound B as Disodium Salt, Foscarnet Sodium Hexahydrate Impurity B as Disodium Salt, Foscarnet Impurity B as Disodium Salt
InChI formel InChI=1S/C3H7O5P.2Na/c1-2-8-9(6,7)3(4)5;;/h2H2,1H3,(H,4,5)(H,6,7);;/q;2*+1/p-2
LEDER [Na+].[Na+].CCOP(=O)([O-])C(=O)[O-]
IUPAC-namn disodium;[ethoxy(oxido)phosphoryl]formate
Molekylvikt (g/mol) 198.02
Kvantitet 25 mg
Formel vikt 197.967 g/mol