Läs mer
Disopyramide 1.0 mg/ml in Methanol, LoGiCal
Produktkod. 30457977
Change view
Kvantitet:
1 ml
Produktkod. Kvantitet
30457977 1 ml
1 options
Produktkod. 30457977 Leverantör LoGiCal Leverantörsnummer LGCAMP0108.0001
LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
Disopyramide 1.0 mg/ml in Methanol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Disopyramide
CAS 9-5-3737
Analyt- eller komponentnamn Disopyramide
Frakt skick Ice Pack (-20°C)
Molekylformel C21 H29 N3 O
Certifikat och efterlevnad ISO 17034
Synonym Disopyramide
InChI formel InChI=1 S/C21H29N3O/c1-16(2)24(17(3)4)15-13-21(20(22)25,18-10-6-5-7-11-18)19-12-8-9-14-23-19/h5-12,14,16-17 H,13,15H2,1-4H3,(H2,22,25)
LEDER CC(C)N(CCC(C(=O)N)(c1ccccc1)c2ccccn2)C(C)C
IUPAC-namn 4-[di(propan-2-yl)amino]-2-phenyl-2-pyridin-2-ylbutanamide
Lösningsmedel eller matris Methanol
Industry Type Forensics
Molekylvikt (g/mol) 339.47
Kvantitet 1 ml
Formel vikt 339.2311 g/mol