Läs mer
Disperse Blue 124, TRC
Produktkod. 30373198
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30373198 100 mg
1 options
Produktkod. 30373198 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD493505100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Disperse Blue 124
CAS 15141-18-1
Molekylformel C16 H19 N5 O4 S
Kvantitet 100 mg
Synonym Ethanol, 2-[N-ethyl-4-[(5-nitro-2-thiazolyl)azo]-m-toluidino]-, acetate (ester) (8CI), Ethanol, 2-[ethyl[3-methyl-4-[(5-nitro-2-thiazolyl)azo]phenyl]amino]-, acetate (ester) (9CI), C.I. 111938, C.I. Disperse Blue 124, Disperse Blue 124, Serisol 3RD, Ethanol, 2-[ethyl[3-methyl-4-[2-(5-nitro-2-thiazolyl)diazenyl]phenyl]amino]-, 1-acetate
InChI formel InChI=1S/C16H19N5O4S/c1-4-20(7-8-25-12(3)22)13-5-6-14(11(2)9-13)18-19-16-17-10-15(26-16)21(23)24/h5-6,9-10H,4,7-8H2,1-3H3
LEDER CCN(CCOC(=O)C)c1ccc(N=Nc2ncc(s2)[N+](=O)[O-])c(C)c1
IUPAC-namn 2-[N-ethyl-3-methyl-4-[(E)-(5-nitro-1,3-thiazol-2-yl)diazenyl]anilino]ethyl acetate
Molekylvikt (g/mol) 377.42
Formel vikt 377.1158