Läs mer
Disperse Blue 60 (Technical Grade), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Disperse Blue 60 |
| CAS | 12217-80-0 |
| Molekylformel | C20H17N3O5 |
| Kvantitet | 250 mg |
| Synonym | 4,11-Diamino-2-(3-methoxypropyl)-1H-naphth[2,3-f]isoindole-1,3,5,10(2H)-tetrone, 2,3-Anthracenedicarboximide, 1,4-diamino-9,10-dihydro-N-(3-methoxypropyl)-9,10-dioxo- (7CI), 2,3-Anthraquinonedicarboximide, 1,4-diamino-N-(3-methoxypropyl)- (6CI), 4,11-Diamino-2-(3-methoxypropyl)-1H-naphth[2,3-f]isoindole-1,3,5,10(2H)-tetrone (ACI), C.I. Disperse Blue 60 (8CI), B 60, C.I. 61104, C.I. Disperse Blue 99, Dianix Turquoise Blue S-BG, Disperse Blue 60, Disperse Blue 99, Disperse Turquoise Blue 60, Disperse Turquoise Blue GL, Disperse Turquoise Blue S-GL, Genacron Brilliant Blue 8G, Kayalon Polyester Turquoise Blue GGF, Kayalon Polyester Turquoise Blue GL-S, Kayalon Polyester Turquoise Blue GL-S 200%, Kayaset B 766, Latyl Brilliant Blue BG, Miketon Polyester Brilliant Blue BG, Palanil Brilliant Blue BG, Palanil Brilliant Blue BG-M, Polycron Turquoise Blue GLP, Qualsperse Blue BNS, Resolin Brilliant Blue BGL, Resolin Brilliant Blue BGL 200, Samaron Blue FFBL, Samaron Brilliant Blue R-GL, Serilene Brilliant Blue 2GN, Sodyecron Brilliant Blue BGE 200, Sumikaron Turquoise Blue G, Sumikaron Turquoise Blue S-GL, Sumikaron Turquoise Blue S-GL 200, Sumikaron Turquoise Blue S-GL double liquid, Sumikaron Turquoise Blue S-GLF, Sumiplast Turquoise Blue G, Synten Blue P-BGFN, Terasil Blue BGE, Terasil Blue BGE 01, Terasil Blue BGE 01-200, Terasil BR Blue BGE 200-017, Terasil Brilliant Blue BGE, Teratop Blue BGE, Tersetile Turquoise GL, TS Turquoise Blue 606, Turquoise Blue S-GL |
| InChI formel | InChI=1S/C20H17N3O5/c1-28-8-4-7-23-19(26)13-14(20(23)27)16(22)12-11(15(13)21)17(24)9-5-2-3-6-10(9)18(12)25/h2-3,5-6H,4,7-8,21-22H2,1H3 |
| LEDER | O=C1C=2C=CC=CC2C(=O)C=3C(N)=C4C(=O)N(C(=O)C4=C(N)C13)CCCOC |
| IUPAC-namn | 4,11-Diamino-2-(3-methoxypropyl)-1H-naphth[2,3-f]isoindole-1,3,5,10(2H)-tetrone |
| Molekylvikt (g/mol) | 379.37 |
| Formel vikt | 379.12 |