Läs mer
Disperse Orange 29 (Technical Grade), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Disperse Orange 29 |
| CAS | 19800-42-1 |
| Molekylformel | C19H15N5O4 |
| Kvantitet | 50 mg |
| Synonym | Phenol, 4-[2-[2-methoxy-4-[2-(4-nitrophenyl)diazenyl]phenyl]diazenyl]-, Phenol, 4-[[2-methoxy-4-[(4-nitrophenyl)azo]phenyl]azo]- (9CI), Phenol, p-[[2-methoxy-4-[(p-nitrophenyl)azo]phenyl]azo]- (8CI), 4-[2-[2-Methoxy-4-[2-(4-nitrophenyl)diazenyl]phenyl]diazenyl]phenol, 4-[[2-Methoxy-4-[(4-nitrophenyl)azo]phenyl]azo]phenol, C.I. Disperse Orange 29, Dianix Yellow Brown SE-R, Disperse Orange 29, Foron Yellow Brown SE-RL, Hisperse Orange C-GS, Intrasil Orange L 2R, Intrasil Orange L 2R200, Palanil Orange GL, Resolin Yellow Brown 3GL, Samaron Yellow Brown HRSL, Sumikaron Orange SE-RBL, Synten Orange P-GRL, Phenol, 4-[2-[2-methoxy-4-[2-(4-nitrophenyl)diazenyl]phenyl]diazenyl]-, Phenol, 4-[[2-methoxy-4-[(4-nitrophenyl)azo]phenyl]azo]- (9CI), Phenol, p-[[2-methoxy-4-[(p-nitrophenyl)azo]phenyl]azo]- (8CI), 4-[2-[2-Methoxy-4-[2-(4-nitrophenyl)diazenyl]phenyl]diazenyl]phenol, 4-[[2-Methoxy-4-[(4-nitrophenyl)azo]phenyl]azo]phenol, C.I. Disperse Orange 29, Dianix Yellow Brown SE-R, Disperse Orange 29, Foron Yellow Brown SE-RL, Hisperse Orange C-GS, Intrasil Orange L 2R, Intrasil Orange L 2R200, Palanil Orange GL, Resolin Yellow Brown 3GL, Samaron Yellow Brown HRSL, Sumikaron Orange SE-RBL, Synten Orange P-GRL |
| InChI formel | InChI=1S/C19H15N5O4/c1-28-19-12-15(22-20-13-2-7-16(8-3-13)24(26)27)6-11-18(19)23-21-14-4-9-17(25)10-5-14/h2-12,25H,1H3 |
| LEDER | COc1cc(ccc1N=Nc2ccc(O)cc2)N=Nc3ccc(cc3)[N+](=O)[O-] |
| IUPAC-namn | 4-[(E)-[2-methoxy-4-[(E)-(4-nitrophenyl)diazenyl]phenyl]diazenyl]phenol |
| Molekylvikt (g/mol) | 377.35 |
| Formel vikt | 377.1124 |