Läs mer
Disperse Orange 37, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Disperse Orange 37 |
| CAS | 13301-61-6 |
| Molekylformel | C17 H15 Cl2 N5 O2 |
| Kvantitet | 1 g |
| Synonym | Propanenitrile, 3-[[4-[(2,6-dichloro-4-nitrophenyl)azo]phenyl]ethylamino]- (9CI), Propionitrile, 3-[p-[(2,6-dichloro-4-nitrophenyl)azo]-N-ethylanilino]- (6CI,8CI), 3-[[4-[2-(2,6-Dichloro-4-nitrophenyl)diazenyl]phenyl]ethylamino]propanenitrile, Akasperse Orange RL 150, C.I. Disperse Orange 37, C.I. Disperse Orange 59, C.I. Disperse Orange 76, Disperse Orange 37, Disperse Orange 59, Disperse Orange 76, Disperse Polyester Yellow Brown, Disperse Yellow Brown 2RCW, Eastman Polyester Dark Orange RL, Eastone Dark Orange R-GLF, Intrasil Dark Orange 3GH, Kayalon Polyester Yellow Brown 3RL, Serilene Yellow Brown 2RL, Serilene Yellow Brown 2RL150 |
| InChI formel | InChI=1S/C17H15Cl2N5O2/c1-2-23(9-3-8-20)13-6-4-12(5-7-13)21-22-17-15(18)10-14(24(25)26)11-16(17)19/h4-7,10-11H,2-3,9H2,1H3 |
| LEDER | CCN(CCC#N)c1ccc(cc1)N=Nc2c(Cl)cc(cc2Cl)[N+](=O)[O-] |
| IUPAC-namn | 3-[4-[(E)-(2,6-dichloro-4-nitrophenyl)diazenyl]-N-ethylanilino]propanenitrile |
| Molekylvikt (g/mol) | 392.24 |
| Formel vikt | 391.0603 |