Learn More
Alfa Aesar™ Disposable IR Cards, 5x10cm, KCl window, 15mm aperture
Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.
Brand:  Alfa Aesar™ 043933.KM
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15496565
  • 3226.01 SEK / Pack of 25
Estimated Shipment date
from Supplier 23-04-2019
Add to basket
Description
Description
  • Size: 5 x 10cm
  • KCl window
  • 15mm aperture
Specifications
Specifications
Disposable IR Cards
25 Pk.