Learn More
Corning™ Dissolvable Microcarriers
Shop All Corning Products
Click to view available options
Kvantitet:
1 g
5 g
10 g
100 g
Unit Size:
Varje
Description
- Offer cell separations that are faster, gentler, and more convenient than other standard commercially available microcarriers
- Comprised of polygalacturonic acid (PGA) polymer chains cross linked via calcium ions pre-coated with either denatured collagen or Corning Synthemax™ II to enhance cell attachment for optimal yield and viability
- Dissolvable within 10 to 20 minutes using EDTA and pectinase for easy separation of cells from the actual microcarriers
- Narrow size distribution (1.02-1.03 g/cm3 density, 200-300μm fully hydrated) and large surface area (5000cm2/g) result in a more homogenous culture
- Optical clarity for visualizing cell morphology
- Presterilized to Sterility Assurance Level (SAL) of 10-6
- Nonpyrogenic
- Applications areas include cell therapy, vaccine production or large-volume adherent cell culture scale-up and cell banking
Specifications
Specifications
| Beskrivning | Dissolvable microcarriers |
| Kulturområde | 5000 cm2/g |
| För användning med (applikation) | Large-volume adherent cell culture scale-up and cell banking |
| Förpackning | Glass Vial |
| Ytbehandling | Corning™ Synthemax II |
| Produkttyp | Dissolvable Microcarrier |
| Material | Polygalacturonic acid (PGA) polymer chains cross linked via calcium ions |
| Partikelstorlek (metrisk) | 200 to 300 μm fully hydrated |
| Kvantitet | 5 g |