Läs mer
Palintest™ Photometer Tube Test
Räckvidd:
0 till 2 mg/l
0 till 20 mg/l
Antal tester:
30
Förpackningsstorlek:
250 tester
Paket med 30
Varje
Beskrivning
- For ultimate convenience, the Palintest™ range of Tubetest reagents offer pre-dispensed reagents for minimal handling with barcoded reagent labelling for ease of use in wastewater applications.
- For a complete wastewater analysis system, use Palintest™ tubetests with any Palintest™ multiparameter photometer and heater block.
- Tubetest reagents are supplied in 16mm tubes for improved resolution, with greater sensitivity in the lower part of the measurement range.
Specifikationer
Specifikationer
| Analyter | Dissolved Oxygen |
| För användning med (utrustning) | Photometer, heater block |
| Antal tester | 30 |
| Kvantitet | 1 Each |
| Räckvidd | 0 till 2 mg/l |
| Typ | Photometer tube test reagent |