Learn More
Dissolved Oxygen Refilling Electrolyte for ECDOGEN-S and ECDOTPII-S DO Electrodes
Eutech™ offers a wide range of Dissolved Oxygen (DO) meters and probes that measure the amount of gaseous oxygen dissolved in a solution in various educational, laboratory and industrial applications.
1550.22 SEK
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
11593765
|
Thermo Scientific Eutech
01X211228 |
DO refilling electrolyte for ECDOGEN-S electrode, 480mL | ECDOGEN-S DO Electrode |
1550.22 SEK
480mL |
Estimated Shipment: 30-04-2024
Log in to see stock. |
||||
11503775
|
Thermo Scientific Eutech
01X211229 |
DO refilling electrolyte for ECDOTPII-S electrode, 480mL | ECDOTPII-S |
1550.22 SEK
480mL |
Estimated Shipment: 30-04-2024
Log in to see stock. |
||||
Description
Eutech™ offers a wide range of Dissolved Oxygen (DO) meters and probes that measure the amount of gaseous oxygen dissolved in a solution in various educational, laboratory and industrial applications. Eutech™ Dissolved Oxygen Refilling Electrolytes are used for ECDOGEN-S and ECDOTPII-S Dissolved Oxygen Process Electrodes.
Features:
- Refill Electrolyte Solution in 480 mL bottles
Specifications
Refilling Electrolyte Solution | |
DO Galvanic Electrode |