Läs mer
Disulfamide, LoGiCal
Produktkod. 30461891
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30461891 10 mg
1 options
Produktkod. 30461891 Leverantör LoGiCal Leverantörsnummer LGCFOR3052.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Disulfamide
CAS 671-88-5
Analyt- eller komponentnamn Disulfamide
Frakt skick Room Temperature
Molekylformel C7 H9 Cl N2 O4 S2
Certifikat och efterlevnad ISO 9001
Synonym 1,3-Benzenedisulfonamide, 4-chloro-6-methyl-, Toluene-2,4-disulfonamide, 5-chloro- (6CI,7CI,8CI), 5-Chloro-2,4-disulfamoyltoluene, 5-Chlorotoluene-2,4-disulfonamide, Desamide, Disamide, Disulfamide, Disulfamide (pharmaceutical), Disulphamide, Diurex, Diurex (Eisai), Diuroblitz, Edemil, Fisiuretik, NSC 38904, Natirene 25, Tolclotidum, Toluidrin, Toluren
InChI formel InChI=1S/C7H9ClN2O4S2/c1-4-2-5(8)7(16(10,13)14)3-6(4)15(9,11)12/h2-3H,1H3,(H2,9,11,12)(H2,10,13,14)
Lösningstyp Neat
LEDER Cc1cc(Cl)c(cc1S(=O)(=O)N)S(=O)(=O)N
IUPAC-namn 4-chloro-6-methylbenzene-1,3-disulfonamide
Industry Type Clinical Testing
Molekylvikt (g/mol) 284.74
Kvantitet 10 mg
Formel vikt 283.9692 g/mol