missing translation for 'onlineSavingsMsg'
Learn More

Dithioerythritol, MP Biomedicals™

Catalog Number p-4403806
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
This item is not returnable. View return policy
6892-68-8
C4H10O2S2
154.24
MFCD00063750
VHJLVAABSRFDPM-ZXZARUISSA-N
dithioerythritol, 1,4-dithioerythritol, dte, 2r,3s-1,4-dimercaptobutane-2,3-diol, erythro-1,4-dimercapto-2,3-butanediol, 2,3-butanediol, 1,4-dimercapto-, 2r,3s-rel, 1,4-dithioerythritol dte, 2r*,3s*-1,4-dimercapto-2,3-butanediol, erythro-2,3-dihydroxy-1,4-butanedithiol, 2r,3s-1,4-disulfanylbutane-2,3-diol
439352
CHEBI:17456
O[C@@H](CS)[C@H](O)CS
This item is not returnable. View return policy
6892-68-8
C4H10O2S2
154.24
MFCD00063750
VHJLVAABSRFDPM-ZXZARUISSA-N
dithioerythritol, 1,4-dithioerythritol, dte, 2r,3s-1,4-dimercaptobutane-2,3-diol, erythro-1,4-dimercapto-2,3-butanediol, 2,3-butanediol, 1,4-dimercapto-, 2r,3s-rel, 1,4-dithioerythritol dte, 2r*,3s*-1,4-dimercapto-2,3-butanediol, erythro-2,3-dihydroxy-1,4-butanedithiol, 2r,3s-1,4-disulfanylbutane-2,3-diol
439352
CHEBI:17456
O[C@@H](CS)[C@H](O)CS
Color White
Assay Percent Range >99%
Quantity 1 g
IUPAC Name (2R,3S)-1,4-disulfanylbutane-2,3-diol
Formula Weight 154.2
Physical Form Crystalline Powder
Chemical Name or Material Dithioerythritol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'