Learn More
Invitrogen™ Dithiothreitol (DTT)
Product Code. 10789174
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
10789174 1 g 1g
1 options
For Research Use Only. All usage must comply with product instructions.
Product Code. 10789174 Supplier Invitrogen™ Supplier No. D1532
 Restricted Item
I lager
Add to basket
Edge
Add to basket
For Research Use Only. All usage must comply with product instructions.
Dithiothreitol (DTT)
Disufide crosslinks of cystines in proteins can be reduced to cysteine residues by dithiothreitol (DTT).
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at room temperature.
Shipping Condition Room Temperature
Form Solid
Quantity 1 g
Product Type Dithiothreitol