Läs mer
Divicine Sulfate, TRC
Produktkod. 30135914
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135914 10 mg
30007135 5 mg
30005078 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135914 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD49445810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5348-48-1
Molekylformel C4H6N4O2 . x(H2O4S)
Synonym 2,6-Diamino-4,5-Pyrimidinediol Sulfate, 2,6-Diamino-3,6-dihydro-4,5-pyrimidinedione Sulfate;
LEDER NC1=NC(=O)C(=C(N)N1)O.NC2=NC(=O)C(=C(N)N2)O.OS(=O)(=O)O
Specifikationer
Kemiskt namn eller material Divicine Sulfate
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
Formel vikt 382.0655
Procent renhet >95%