Learn More
Divinylbenzene, 55%, mixture of isomers, stab. with 1000ppm 4-tert-butylcatechol, Thermo Scientific Chemicals
Description
Used as a co-monomer in the production of ion exchange beads and gel chromatography. As a monomer in the production of synthetic rubber and casting resins. Also used in polymeric core shell nanoparticles and as a crosslinking agent. It may be used as a monomer for the synthesis of highly crosslinked macroporous styrene/DVB polymers. It may also be used to synthesize monodispersed poly-DVB microspheres.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 0.912 g/mL |
Flash Point | 62°C (143°F) |
Refractive Index | 1.574 |
Quantity | 50 g |
Solubility Information | Insoluble in water. |
Formula Weight | 130.19 |
Percent Purity | 55% |
Physical Form | Liquid |
Chemical Name or Material | Divinylbenzene, Mixture of isomers, Stabilized with 1000ppm 4-tert-butylcatechol |
RUO – Research Use Only