missing translation for 'onlineSavingsMsg'
Learn More

DKC1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25420-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16883995

  • 1236.20 SEK / 20µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

May function as a transcriptional repressor. May also function as a ubiquitin ligase mediating ubiquitination of chromatin-associated TP53. Functions in cell survival and proliferation through control of the cell cycle. Functions in the p53 and pRB tumor suppressor pathways and regulates the cyclin CCNA2 transcription.; FUNCTION: Identified as a cellular target of the adenoviral oncoprotein E1A, it is required for both transcriptional activation and repression of viral genes.
Specifications

Specifications

DKC1
0.39 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O60832
CBF5, CBF5 homolog, DKC, DKC1, Dyskerin, NAP57, NOLA4, Nucleolar protein NAP57, snoRNP protein DKC1, XAP101
Rabbit
IgG
20 μL
-20°C
Primary
1736
Western Blot
Unconjugated
DKC1
Liquid
DKC1
DKC1 Fusion Protein Ag7992
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers