Läs mer
DKK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
4652.90 SEK
Specifikationer
| Antigen | DKK1 |
|---|---|
| Koncentration | 0.23 mg/mL |
| Användningsområden | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Klassificering | Polyclonal |
| Konjugera | Unconjugated |
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|
16880715
|
Proteintech
21112-1-AP-150UL |
150 μL |
4652.90 SEK
150µL |
Vänligen Logga in för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag! | |||||
Beskrivning
This gene encodes a protein that is a member of the dickkopf family. It is a secreted protein with two cysteine rich regions and is involved in embryonic development through its inhibition of the WNT signaling pathway. Elevated levels of DKK1 in bone marrow plasma and peripheral blood is associated with the presence of osteolytic bone lesions in patients with multiple myeloma.Specifikationer
| DKK1 | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O54908, O94907 | |
| 13380, 22943, 293897 | |
| DKK1 Fusion Protein Ag15110 | |
| Primary | |
| -20°C | |
| DKK1 |
| 0.23 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Dickkopf 1, Dickkopf related protein 1, DKK 1, DKK1, hDkk 1, SK | |
| Dkk1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |