missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DKK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21051-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that is a member of the dickkopf family. The secreted protein contains two cysteine rich regions and is involved in embryonic development through its interactions with the Wnt signaling pathway. It can act as either an agonist or antagonist of Wnt/beta-catenin signaling, depending on the cellular context and the presence of the co-factor kremen 2. Activity of this protein is also modulated by binding to the Wnt co-receptor LDL-receptor related protein 6.Specifications
DKK2 | |
Polyclonal | |
Unconjugated | |
DKK2 | |
Dickkopf 2, Dickkopf related protein 2, DKK 2, DKK2, hDkk 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27123, 295445, 56811 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9QYZ8, Q9UBU2 | |
DKK2 | |
DKK2 Fusion Protein Ag12405 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |