missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DKK4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27080-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that is a member of the dickkopf family. The secreted protein contains two cysteine rich regions and is involved in embryonic development through its interactions with the Wnt signaling pathway. Activity of this protein is modulated by binding to the Wnt co-receptor and the co-factor kremen 2.Specifications
DKK4 | |
Polyclonal | |
Unconjugated | |
DKK4 | |
Dickkopf 4, Dickkopf related protein 4, DKK 4, DKK4, hDkk 4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
234130, 27121 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8VEJ3, Q9UBT3 | |
DKK4 | |
DKK4 Fusion Protein Ag24209 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |