missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DKK4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | DKK4 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16825375
|
Proteintech
27080-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16815375
|
Proteintech
27080-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein that is a member of the dickkopf family. The secreted protein contains two cysteine rich regions and is involved in embryonic development through its interactions with the Wnt signaling pathway. Activity of this protein is modulated by binding to the Wnt co-receptor and the co-factor kremen 2.Specifications
DKK4 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q8VEJ3, Q9UBT3 | |
234130, 27121 | |
DKK4 Fusion Protein Ag24209 | |
Primary | |
-20°C | |
DKK4 |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Dickkopf 4, Dickkopf related protein 4, DKK 4, DKK4, hDkk 4 | |
DKK4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |