Läs mer
DL-2-Aminoadipic acid, 99%, anhydrous
Produktkod. 11965381 Handla allt Thermo Scientific Chemicals Produkter
Change view
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11965381 5 g 5g
11955381 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11965381 Leverantör Thermo Scientific Acros Leverantörsnummer 441790050
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
CAS: 542-32-5 | C6H11NO4 | 161.16 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 542-32-5
Molekylformel C6H11NO4
Molekylvikt (g/mol) 161.16
MDL-nummer MFCD00063119,MFCD00063118
InChI-nyckel OYIFNHCXNCRBQI-UHFFFAOYNA-N
Synonym dl-2-aminoadipic acid, aminoadipic acid, alpha-aminoadipic acid, 2-aminoadipic acid, dl-alpha-aminoadipic acid, 2-aminoadipate, alpha-aminoadipate, hexanedioic acid, 2-amino, 2-amino-hexanedioic acid, dl-2-aminohexanedioic acid
PubChem CID 469
ChEBI CHEBI:37024
IUPAC-namn 2-aminohexanedioic acid
LEDER NC(CCCC(O)=O)C(O)=O
Specifikationer
Kemiskt namn eller material DL-2-Aminoadipic acid
Smältpunkt 176.0°C to 177.0°C
Färg White
Infrarött spektrum Authentic
Analysprocentintervall 99%
Kvantitet 5 g
Merck Index 15,41
Löslighetsinformation Solubility in water: .. Other solubilities: sparingly soluble in alcohol and ether
Formel vikt 161.16
Procent renhet 99%
Fysisk form Crystalline Powder or Powder