Läs mer
DL-2-Aminobutyric Acid Methyl Ester Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | DL-2-Aminobutyric Acid Methyl Ester Hydrochloride |
| CAS | 7682-18-0 |
| Molekylformel | C5H12ClNO2 |
| Kvantitet | 10 g |
| Synonym | 2-Aminobutanoic Acid Methyl Ester Hydrochloride, DL-Methyl 2-Aminobutanoate Hydrochloride, DL-alpha-Aminobutyric alphacid μethyl epsilonster etaydrochloride, Methyl 2-alphaminobutanoate etaydrochloride, Methyl 2-alphaminobutyrate etaydrochloride, Methyl DL-2-alphaminobutyrate hydrochloride, Methyl alpha-alphaminobutyrate etaydrochloride |
| InChI formel | InChI=1S/C5H11NO2.ClH/c1-3-4(6)5(7)8-2;/h4H,3,6H2,1-2H3;1H |
| LEDER | Cl.CCC(N)C(=O)OC |
| IUPAC-namn | methyl 2-aminobutanoate;hydrochloride |
| Molekylvikt (g/mol) | 153.61 |
| Formel vikt | 153.0557 |