Läs mer
DL-2-Aminocaprylic Acid, TRC
Produktkod. 30323717
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30323717 5 g
1 options
Produktkod. 30323717 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6031855G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material DL-2-Aminocaprylic Acid
CAS 644-90-6
Molekylformel C8H17NO2
Kvantitet 5 g
Synonym 2-Amino-Octanoic Acid, (±)-2-Amino-Octanoic Acid, (±)-2-Aminocaprylic Acid, (±)-2-Aminooctanoic Acid, (±)-alpha-Aminocaprylic Acid, 2-Amino-DL-caprylic Acid, 2-Aminooctanoic Acid, DL-2-Aminocaprylic Acid, DL-2-Aminooctanoic Acid, DL-alpha-Aminocaprylic Acid, DL-alpha-Aminooctanoic Acid, NSC 123023, NSC 20147, alpha-Aminocaprylic Acid, alpha-Aminooctanoic Acid
InChI formel InChI=1S/C8H17NO2/c1-2-3-4-5-6-7(9)8(10)11/h7H,2-6,9H2,1H3,(H,10,11)
LEDER CCCCCCC(N)C(=O)O
IUPAC-namn 2-aminooctanoic acid
Molekylvikt (g/mol) 159.23
Formel vikt 159.1259