Läs mer
DL-2-Phenylglycinamide, TRC
Produktkod. 30411049
Change view
Kvantitet:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30411049 50 mg
30425158 1 g
30409738 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30411049 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP32710550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 700-63-0
Molekylformel C8H10N2O
Molekylvikt (g/mol) 150.18
Synonym (±)-alpha-Aminobenzeneacetamide, 2-Amino-2-phenylacetamide, DL-Phenylglycinamide, DL-Phenylglycine Amide, DL-alpha-Phenylglycine Amide, NSC 47695, alpha-Amino-alpha-phenylacetamide
IUPAC-namn 2-amino-2-phenylacetamide
LEDER NC(C(=O)N)c1ccccc1
Specifikationer
Kemiskt namn eller material DL-2-Phenylglycinamide
Kvantitet 50 mg
InChI formel InChI=1S/C8H10N2O/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7H,9H2,(H2,10,11)
Formel vikt 150.0793