Learn More
DL-2-Phenylpropionic acid, 98%, Thermo Scientific Chemicals
Catalog Number p-3779442
474.95 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
492-37-5
MFCD00002650
YPGCWEMNNLXISK-UHFFFAOYSA-N
2-phenylpropionic acid, hydratropic acid, dl-2-phenylpropionic acid, alpha-methylphenylacetic acid, 2-phenyl-propionic acid, +/--2-phenylpropionic acid, alpha-methylbenzeneacetic acid, benzeneacetic acid, .alpha.-methyl, hydratropasaeure, alpha-phenylpropionic acid
10296
CHEBI:48526
2-phenylpropanoic acid
CC(C1=CC=CC=C1)C(=O)O
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
492-37-5
MFCD00002650
YPGCWEMNNLXISK-UHFFFAOYSA-N
2-phenylpropionic acid, hydratropic acid, dl-2-phenylpropionic acid, alpha-methylphenylacetic acid, 2-phenyl-propionic acid, +/--2-phenylpropionic acid, alpha-methylbenzeneacetic acid, benzeneacetic acid, .alpha.-methyl, hydratropasaeure, alpha-phenylpropionic acid
10296
CHEBI:48526
2-phenylpropanoic acid
CC(C1=CC=CC=C1)C(=O)O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 5°C
Color Yellow
Density 1.1000g/mL
Boiling Point 260°C to 262°C
Flash Point 148°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C9H10O2
Linear Formula CH3CH(C6H5)CO2H
Refractive Index 1.5215 to 1.5235
Quantity 10 g
Beilstein 09, 524
Specific Gravity 1.1
Solubility Information Solubility in water: 10g/L (20°C)
Molecular Weight (g/mol) 150.18
Viscosity 44 mPa.s (20°C)
Formula Weight 150.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material DL-2-Phenylpropionic acid
Show More Show Less
compliance-icons
Product Identifier
  • DL-2-Phenylpropionic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.