Läs mer
DL-3-Phenylserine Hydrate, TRC
Produktkod. 30422423
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30422423 1 g
1 options
Produktkod. 30422423 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP3193851G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material DL-3-Phenylserine Hydrate
CAS 207605-47-8
Molekylformel C9H11NO3 . xH 2O
Kvantitet 1 g
Synonym β-Hydroxy-phenylalanine Hydrate
InChI formel InChI=1S/C9H11NO3.H2O/c10-7(9(12)13)8(11)6-4-2-1-3-5-6;/h1-5,7-8,11H,10H2,(H,12,13);1H2
LEDER O=C(C(C(C1=CC=CC=C1)O)N)O.O
IUPAC-namn 2-amino-3-hydroxy-3-phenylpropanoic acid hydrate
Molekylvikt (g/mol) 181.19