missing translation for 'onlineSavingsMsg'
Learn More

DL-6-Methyl-5-hepten-2-ol, 99%, Thermo Scientific Chemicals

Catalog Number p-3758406
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
4630-06-2
C8H16O
128.21
MFCD00004561
OHEFFKYYKJVVOX-UHFFFAOYNA-N
CC(O)CCC=C(C)C
This item is not returnable. View return policy
4630-06-2
C8H16O
128.21
MFCD00004561
OHEFFKYYKJVVOX-UHFFFAOYNA-N
CC(O)CCC=C(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.8400g/mL
Boiling Point 78.0°C (14.0 mmHg)
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)2C=CHCH2CH2CH(OH)CH3
Refractive Index 1.4475 to 1.4495
Quantity 25 mL
Beilstein 01, IV, 2167
Specific Gravity 0.84
Synonym 6-methyl-5-hepten-2-ol, sulcatol, 5-hepten-2-ol, 6-methyl, r-sulcatol, 2-methyl-2-hepten-6-ol, +/--6-methyl-5-hepten-2-ol, 6-methyl-hept-5-en-2-ol, r---6-methyl-5-hepten-2-ol, 6-hydroxy-2-methyl-2-heptene, acmc-20apkw
IUPAC Name 6-methylhept-5-en-2-ol
PubChem CID 20745
ChEBI CHEBI:15833
Formula Weight 128.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material DL-6-Methyl-5-hepten-2-ol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'