Läs mer
DL-alpha-(Difluoromethyl)arginine, TRC
Produktkod. 30133004
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133004 10 mg
17908324 5 mg
17957354 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133004 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44584010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 69955-43-7
Molekylformel C7 H14 F2 N4 O2
Molekylvikt (g/mol) 224.21
Synonym 2-(Difluoromethyl)arginine, alpha-(Difluoromethyl)-DL-arginine, alpha-Difluoromethylarginine, DFMA, RMI 71897;
IUPAC-namn 2-amino-5-carbamimidamido-2-(difluoromethyl)pentanoic acid
LEDER NC(=N)NCCCC(N)(C(F)F)C(=O)O
Specifikationer
Kemiskt namn eller material DL-a-(Difluoromethyl)arginine
InChI formel InChI=1S/C7H14F2N4O2/c8-4(9)7(12,5(14)15)2-1-3-13-6(10)11/h4H,1-3,12H2,(H,14,15)(H4,10,11,13)
Kvantitet 10 mg
Formel vikt 224.1085