missing translation for 'onlineSavingsMsg'
Learn More

DL-α-Bromophenylacetic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-4258812
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4870-65-9
C8H7BrO2
215.04
WAKFRZBXTKUFIW-UHFFFAOYSA-N
alpha-bromophenylacetic acid, bromo phenyl acetic acid, .alpha.-bromophenylacetic acid, bromophenylacetic acid, bromo-phenyl-acetic acid, bromo-phenylacetic acid, dl-.alpha.-bromophenylacetic acid, benzeneacetic acid, .alpha.-bromo, phenylbromoacetic acid, bromo phenyl aceticacid
97919
2-bromo-2-phenylacetic acid
C1=CC=C(C=C1)C(C(=O)O)Br
This item is not returnable. View return policy
4870-65-9
C8H7BrO2
215.04
WAKFRZBXTKUFIW-UHFFFAOYSA-N
alpha-bromophenylacetic acid, bromo phenyl acetic acid, .alpha.-bromophenylacetic acid, bromophenylacetic acid, bromo-phenyl-acetic acid, bromo-phenylacetic acid, dl-.alpha.-bromophenylacetic acid, benzeneacetic acid, .alpha.-bromo, phenylbromoacetic acid, bromo phenyl aceticacid
97919
2-bromo-2-phenylacetic acid
C1=CC=C(C=C1)C(C(=O)O)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 82.0°C to 83.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH(Br)CO2H
Quantity 5 g
Beilstein 09,451
Solubility Information Solubility in water: insoluble
Formula Weight 215.04
Percent Purity 97%
Chemical Name or Material DL-α-Bromophenylacetic acid
compliance-icons
Product Identifier
  • DL-alpha-Bromophenylacetic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'