missing translation for 'onlineSavingsMsg'
Läs mer

dl-Camphor, Sigma-Aldrich™

Produktkod. 15876780
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 g
Förpackningsstorlek:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15876780 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
MQ200 Graded - For Research and Non-regulated Industrial Applications
Produktkod. 15876780 Leverantör Sigma Aldrich Leverantörsnummer 8414560250
 Begränsad artikel
Beräknad leverans: 18-11-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy
MQ200 Graded - For Research and Non-regulated Industrial Applications

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 76-22-2
Smältpunkt 180°C
Densitet 0.99 g/cm3 at 20°C
Kokpunkt 205°C/1013 hPa
MDL-nummer MFCD00074738
Kvantitet 250 g
Synonym DL-Camphor, DL-Camphor
Molekylvikt (g/mol) 152.23
Kvalitet Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.