Läs mer
DL-Ethyl Lactate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Ethyl lactate |
| CAS | 97-64-3 |
| Molekylformel | C5H10O3 |
| Kvantitet | 100 g |
| Synonym | Propanoic acid, 2-hydroxy-, ethyl ester, Lactic acid, ethyl ester (6CI,8CI), (±)-Ethyl 2-hydroxypropionate, (±)-Ethyl lactate, (±)-Lactic acid ethyl ester, 2-Hydroxypropanoic acid ethyl ester, Actylol, Acytol, DL-Ethyl lactate, Ethyl 2-hydroxypropanoate, Ethyl 2-hydroxypropionate, Ethyl lactate, Ethyl rac-lactate, Ethyl α-hydroxypropionate, NSC 8850, PBR 40, Purasolv EL, Purasolv ELS, Solactol, Vertec ELS, VertecBio EL, dl-Lactic acid ethyl ester |
| InChI formel | InChI=1S/C5H10O3/c1-3-8-5(7)4(2)6/h4,6H,3H2,1-2H3 |
| LEDER | CCOC(=O)C(C)O |
| IUPAC-namn | ethyl 2-hydroxypropanoate |
| Molekylvikt (g/mol) | 118.13 |
| Formel vikt | 118.063 |