Läs mer
DL-Glyceraldehyde, TRC
Produktkod. 30136536
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136536 100 mg
30136194 500 mg
30010895 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136536 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG598215100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 56-82-6
Molekylformel C3H6O3
Molekylvikt (g/mol) 90.08
Synonym (±)-2,3-Dihydroxy-propanal, (±)-Glyceraldehyde, 2,3-Dihydroxypropanal, 2,3-Dihydroxypropionaldehyde, DL-Glyceric aldehyde, Glyceraldehyde, Glyceric aldehyde, Glycerinaldehyde, Glycerose, NSC 67934, dl-Glyceraldehyde, alpha,beta-Dihydroxypropionaldehyde
IUPAC-namn 2,3-dihydroxypropanal
LEDER OCC(O)C=O
Specifikationer
Kemiskt namn eller material DL-Glyceraldehyde
Kvantitet 100 mg
InChI formel InChI=1S/C3H6O3/c4-1-3(6)2-5/h1,3,5-6H,2H2
Formel vikt 90.0317