missing translation for 'onlineSavingsMsg'
Learn More

DL-Isoleucine, ≥99%, MP Biomedicals™

325.47 SEK - 2071.62 SEK

Chemical Identifiers

CAS 443-79-8
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.175
MDL Number MFCD00004268
InChI Key AGPKZVBTJJNPAG-CRCLSJGQSA-N
Synonym d-alloisoleucine, d-allo-isoleucine, 2r,3s-2-amino-3-methylpentanoic acid, allo-d-isoleucine, alloisoleucine, d, h-d-allo-ile-oh, threo-d-isoleucine, dl-allo-isoleucine, hile, h-dl-allo-ile-oh
PubChem CID 94206
ChEBI CHEBI:20899
IUPAC Name (2R,3S)-2-amino-3-methylpentanoic acid
SMILES CCC(C)C(C(=O)O)N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11338929 View Documents MP Biomedicals
0210208210
10g 325.47 SEK
10g View alternative products
Save Up to 
»
Estimated Shipment
30-09-2019
Add to basket
 
11368929 View Documents MP Biomedicals
0210208280
100g 2071.62 SEK
100g View alternative products
Save Up to 
»
Estimated Shipment
19-09-2019
Add to basket
 
11348929 View Documents MP Biomedicals
0210208225
25g This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
N/A View alternative products
Save Up to 
»
Specifications

Chemical Identifiers

443-79-8
131.175
AGPKZVBTJJNPAG-CRCLSJGQSA-N
94206
(2R,3S)-2-amino-3-methylpentanoic acid
C6H13NO2
MFCD00004268
d-alloisoleucine, d-allo-isoleucine, 2r,3s-2-amino-3-methylpentanoic acid, allo-d-isoleucine, alloisoleucine, d, h-d-allo-ile-oh, threo-d-isoleucine, dl-allo-isoleucine, hile, h-dl-allo-ile-oh
CHEBI:20899
CCC(C)C(C(=O)O)N

Specifications

DL-Isoleucine
≥99%
MFCD00004268
AGPKZVBTJJNPAG-CRCLSJGQSA-N
(2R,3S)-2-amino-3-methylpentanoic acid
94206
131.17
0.991
10g
443-79-8
C6H13NO2
d-alloisoleucine, d-allo-isoleucine, 2r,3s-2-amino-3-methylpentanoic acid, allo-d-isoleucine, alloisoleucine, d, h-d-allo-ile-oh, threo-d-isoleucine, dl-allo-isoleucine, hile, h-dl-allo-ile-oh
CCC(C)C(C(=O)O)N
131.175
CHEBI:20899
Powder
White