Läs mer
DL-Lauroylcarnitine Chloride, TRC
Produktkod. 30468060
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30468060 50 mg
1 options
Produktkod. 30468060 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL18472050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material DL-Lauroylcarnitine Chloride
CAS 14919-37-0
Molekylformel C19 H38 N O4 . Cl
Kvantitet 50 mg
Synonym (3-Carboxy-2-hydroxypropyl)trimethylammonium Chloride Laurate, 3-Carboxy-N,N,N-trimethyl-2-[(1-oxododecyl)oxy]-1-propanaminium Chloride, (3-Carboxy-2-hydroxypropyl)trimethylammonium Chloride Laurate (Ester), 3-Carboxy-2-(dodecanoyloxy)-N,N,N-trimethylpropan-1-aminium Chloride
InChI formel InChI=1S/C19H37NO4.ClH/c1-5-6-7-8-9-10-11-12-13-14-19(23)24-17(15-18(21)22)16-20(2,3)4;/h17H,5-16H2,1-4H3;1H
LEDER O=C(CCCCCCCCCCC)OC(C[N+](C)(C)C)CC(O)=O.[Cl-]
Molekylvikt (g/mol) 379.96
Formel vikt 379.2489