missing translation for 'onlineSavingsMsg'
Learn More

DL-Phenylalaninol, 95%, Thermo Scientific Chemicals

Catalog Number 15450319
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
16088-07-6
C9H13NO
151.209
MFCD00066689
STVVMTBJNDTZBF-UHFFFAOYSA-N
dl-phenylalaninol, 2-amino-3-phenyl-1-propanol, phenylalaninol, s-phenylalaninol, l-2-amino-3-phenyl-1-propanol, s-2-benzylethanolamine, benzenepropanol, beta-amino, s-beta-aminobenzenepropanol, chembl21676, 2-amino-3-phenyl-propan-1-ol
76652
2-amino-3-phenylpropan-1-ol
C1=CC=C(C=C1)CC(CO)N
This item is not returnable. View return policy
16088-07-6
C9H13NO
151.209
MFCD00066689
STVVMTBJNDTZBF-UHFFFAOYSA-N
dl-phenylalaninol, 2-amino-3-phenyl-1-propanol, phenylalaninol, s-phenylalaninol, l-2-amino-3-phenyl-1-propanol, s-2-benzylethanolamine, benzenepropanol, beta-amino, s-beta-aminobenzenepropanol, chembl21676, 2-amino-3-phenyl-propan-1-ol
76652
2-amino-3-phenylpropan-1-ol
C1=CC=C(C=C1)CC(CO)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 69°C to 73°C
UN Number UN3259
Beilstein 2208237
Formula Weight 151.21
Percent Purity 96%
Quantity 5 g
Physical Form Solid
Chemical Name or Material DL-Phenylalaninol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'