missing translation for 'onlineSavingsMsg'
Learn More

DL-Phenylglycinol, 95%, Thermo Scientific Chemicals

Catalog Number p-7055370
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
7568-92-5
C8H11NO
137.18
MFCD00130145
IJXJGQCXFSSHNL-UHFFFAOYNA-N
2-phenylglycinol, dl-2-phenylglycinol, dl-phenylglycinol, 2-amino-2-phenylethan-1-ol, 2-phenyl-dl-glycinol, beta-aminophenethyl alcohol, 2-amino-2-phenyl-ethanol, benzeneethanol, beta-amino, r-2-phenylglycinol, phenethyl alcohol, beta-amino
92466
2-amino-2-phenylethanol
NC(CO)C1=CC=CC=C1
This item is not returnable. View return policy
7568-92-5
C8H11NO
137.18
MFCD00130145
IJXJGQCXFSSHNL-UHFFFAOYNA-N
2-phenylglycinol, dl-2-phenylglycinol, dl-phenylglycinol, 2-amino-2-phenylethan-1-ol, 2-phenyl-dl-glycinol, beta-aminophenethyl alcohol, 2-amino-2-phenyl-ethanol, benzeneethanol, beta-amino, r-2-phenylglycinol, phenethyl alcohol, beta-amino
92466
2-amino-2-phenylethanol
NC(CO)C1=CC=CC=C1

A chiral arylalkylamine used as organocatalysts. Used in the synthesis of chiral diamines also in determination of chiral recognition properties of novel fluorescent chemosensors for amino acid. an efficient and practical synthesis of l-. alpha.-amino acids using (r)-phenylglycinol as a chiral auxiliary.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A chiral arylalkylamine used as organocatalysts. Used in the synthesis of chiral diamines also in determination of chiral recognition properties of novel fluorescent chemosensors for amino acid. an efficient and practical synthesis of l-. alpha.-amino acids using (r)-phenylglycinol as a chiral auxiliary.

Solubility
Soluble in chloroform, methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 49°C to 53°C
Color White
Beilstein 971219
Solubility Information Soluble in chloroform,methanol.
Formula Weight 137.18
Percent Purity 95%
Quantity 5 g
Physical Form Solid
Chemical Name or Material DL-Phenylglycinol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'