Läs mer
DL-Tetrahydropalmatine, TRC
Produktkod. 30409076
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30409076 1 g
30412511 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30409076 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT2942001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2934-97-6
Molekylformel C21 H25 N O4
Molekylvikt (g/mol) 355.43
Synonym 6H-Dibenzo[a,g]quinolizine, 5,8,13,13a-tetrahydro-2,3,9,10-tetramethoxy-, 6H-Dibenzo[a,g]quinolizine, 5,8,13,13a-tetrahydro-2,3,9,10-tetramethoxy-, (±)-, Berbine, 2,3,9,10-tetramethoxy-, (±)- (8CI), DL-Berbine, 2,3,9,10-tetramethoxy- (6CI), (R,S)-Tetrahydropalmatine, (±)-Corydalis B, (±)-Tetrahydropalmatine, 5,8,13,13a-Tetrahydro-2,3,9,10-tetramethoxy-6H-dibenzo[a,g]quinolizine, DL-Tetrahydropalmatine, THP, Tetrahydropalmatine, dl-Tetrahydropalmatine
IUPAC-namn 2,3,9,10-tetramethoxy-6,8,13,13a-tetrahydro-5H-isoquinolino[3,2-a]isoquinoline
LEDER COc1cc2CCN3Cc4c(CC3c2cc1OC)ccc(OC)c4OC
Specifikationer
Kemiskt namn eller material DL-Tetrahydropalmatine
Kvantitet 1 g
InChI formel InChI=1S/C21H25NO4/c1-23-18-6-5-13-9-17-15-11-20(25-3)19(24-2)10-14(15)7-8-22(17)12-16(13)21(18)26-4/h5-6,10-11,17H,7-9,12H2,1-4H3
Formel vikt 355.1784