Läs mer
DL-threo-beta-(3,4-Methylenedioxyphenyl)serine Acetate Salt, TRC
Produktkod. 30469201
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30469201 250 mg
1 options
Produktkod. 30469201 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM304025250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material DL-threo-beta-(3,4-Methylenedioxyphenyl)serine Acetate Salt
CAS 109910-66-9
Molekylformel C12H15NO7
Kvantitet 250 mg
Synonym (R,S)-rel-α-Amino-β-hydroxy-1,3-benzodioxole-5-propanoic Acid Acetate Salt
InChI formel InChI=1S/C10H11NO5.C2H4O2/c11-8(10(13)14)9(12)5-1-2-6-7(3-5)16-4-15-6;1-2(3)4/h1-3,8-9,12H,4,11H2,(H,13,14);1H3,(H,3,4)/t8-,9+;/m1./s1
LEDER CC(=O)O.C1OC2=C(O1)C=C(C=C2)[C@@H]([C@H](C(=O)O)N)O
IUPAC-namn acetic acid;(2R,3S)-2-amino-3-(1,3-benzodioxol-5-yl)-3-hydroxypropanoic acid
Molekylvikt (g/mol) 285.25
Formel vikt 285.0849