Läs mer
DL-threo-Beta-Phenylserine, TRC
Produktkod. 30342785
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30342785 5 mg
1 options
Produktkod. 30342785 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB4061785MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material DL-threo-beta-Phenylserine
CAS 7695-56-9
Molekylformel C9 H11 N O3
Kvantitet 5 mg
Synonym 2-Amino-3-hydroxy-3-phenylpropanoic Acid, rel-(βS)-β-Hydroxy-D-phenylalanine, DL-threo-3-Phenylserine
InChI formel InChI=1S/C9H11NO3/c10-7(9(12)13)8(11)6-4-2-1-3-5-6/h1-5,7-8,11H,10H2,(H,12,13)
LEDER NC(C(O)c1ccccc1)C(=O)O
IUPAC-namn 2-amino-3-hydroxy-3-phenyl-propanoic acid
Molekylvikt (g/mol) 181.19
Formel vikt 181.0739