Läs mer
DL-threo-Droxidopa, TRC
Produktkod. 30382191
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30382191 10 mg
1 options
Produktkod. 30382191 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD68995010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material DL-threo-Droxidopa
CAS 3916-18-5
Molekylformel C9 H11 N O5
Kvantitet 10 mg
Synonym D-Tyrosine, β,3-dihydroxy-, (βS)-rel-, DL-Tyrosine, β,3-dihydroxy-, threo-, Serine, 3-(3,4-dihydroxyphenyl)-, DL-threo- (8CI), rel-(βS)-β,3-Dihydroxy-D-tyrosine, DL-threo-3,4-Dihydroxyphenylserine, DL-threo-DOPS, DL-threo-Dihydroxyphenylserine, threo-2-Amino-3-(3,4-dihydroxyphenyl)-3-hydroxypropionic acid, DL-threo-Droxidopa
InChI formel InChI=1S/C9H11NO5/c10-7(9(14)15)8(13)4-1-2-5(11)6(12)3-4/h1-3,7-8,11-13H,10H2,(H,14,15)/t7-,8+/m0/s1
LEDER N[C@@H]([C@H](O)c1ccc(O)c(O)c1)C(=O)O
IUPAC-namn (2S,3R)-2-amino-3-(3,4-dihydroxyphenyl)-3-hydroxypropanoic acid
Molekylvikt (g/mol) 213.19
Formel vikt 213.0637