Learn More
DLGAP3 Monoclonal Antibody (OTI1D3), TrueMAB™, OriGene
Product Code. 17555554
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17555554 30 μL 30µL
17535554 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17555554 Supplier Origene Technologies Supplier No. TA811498S
 Restricted Item
Beräknad leverans: 27-11-2026
Mouse Monoclonal Antibody
DLGAP3 Monoclonal antibody specifically detects DLGAP3 in Human samples. It is validated for Western Blot
Specifications
Antigen DLGAP3
Applications Western Blot
Classification Monoclonal
Clone OTI1D3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CYP26B1
Gene Accession No. Q9NR63
Gene Alias BC058433; DAP3; DAP-3; discs large homolog associated protein 3; discs, large (Drosophila) homolog-associated protein 3; disks large-associated protein 3; DLG associated protein 3; Dlgap3; Prpl8; PSD-95/SAP90-associated protein-3; PSD-95/SAP90-binding protein 3; SAP90/PSD 95 associated protein 3; SAP90/PSD-95-associated protein 3; SAPAP3
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 229-512 of human CYP26B1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56603
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1