missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ DLST Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551794
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence: QNTCAMLTTF NEIDMSNIQE MRARHKEAFL KKHNLKLGFM SAFVKASAFA LQEQPVVNAV IDDTTKEVVY RDYIDISVAV ATPRGLVVPV IRNVEAMNFA DIERTITELG EKARKNELAI EDMDGGTFTI SNGGVFGSLF GTPII Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
This gene encodes a mitochondrial protein that belongs to the 2-oxoacid dehydrogenase family. This protein is one of the three components (the E2 component) of the 2-oxoglutarate dehydrogenase complex that catalyzes the overall conversion of 2-oxoglutarate to succinyl-CoA and CO(2). Alternatively spliced transcript variants have been found for this gene.
Specifikationer
| DLST | |
| Polyclonal | |
| Unconjugated | |
| Dlst | |
| 1600017E01Rik; 2-oxoglutarate dehydrogenase complex component E2; 4632413C10Rik; 4930529O08Rik; dihydrolipoamide S-succinyltransferase; dihydrolipoamide S-succinyltransferase (E2 component of 2-oxo-glutarate complex); dihydrolipoamide succinyltransferase component of 2-oxoglutarate dehydrogenase complex; Dihydrolipoyllysine-residue succinyltransferase; dihydrolipoyllysine-residue succinyltransferase component of 2-oxoglutarate dehydrogenase complex, mitochondrial; DLST; DLTS; E2; E2K; OGDC-E2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1743, 299201, 78920 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P36957, Q01205, Q9D2G2 | |
| Dlst | |
| Recombinant protein corresponding to Human DLST. Recombinant protein control fragment (Product #RP-88908). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering