Läs mer
DMAB-anabaseine Dihydrochloride, TRC
Produktkod. 30004337
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004337 10 mg
30010569 1 mg
30003630 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30004337 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD67840810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 154149-38-9
Molekylformel C19 H21 N3 . 2[H Cl]
Molekylvikt (g/mol) 364.31
Synonym 4-[(5,6-Dihydro[2,3'-bipyridin]-3(4H)-ylidene)methyl]-N,N-dimethylbenzenamine Dihydrochloride;
IUPAC-namn N,N-dimethyl-4-[(E)-[6-(3-pyridyl)-3,4-dihydro-2H-pyridin-5-ylidene]methyl]aniline;dihydrochloride
LEDER Cl.Cl.CN(C)c1ccc(\C=C\2/CCCN=C2c3cccnc3)cc1
Specifikationer
Kemiskt namn eller material DMAB-anabaseine Dihydrochloride
Kvantitet 10 mg
InChI formel InChI=1S/C19H21N3.2ClH/c1-22(2)18-9-7-15(8-10-18)13-16-5-4-12-21-19(16)17-6-3-11-20-14-17;;/h3,6-11,13-14H,4-5,12H2,1-2H3;2*1H/b16-13+;;
Formel vikt 363.1269