Läs mer
DMAT, MedChemExpress
Produktkod. 16472697
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16472697 50 mg
16456608 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16472697 Leverantör Medchem Express Leverantörsnummer HY15535
MedChemExpress DMAT is a potent and specific CK2 inhibitor with an IC50 value of 130 nM.
DMAT is a potent and specific CK2 inhibitor with an IC50 value of 130 nM.
Specifikationer
Kemiskt namn eller material DMAT
CAS 749234-11-5
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C9H7Br4N3
Kvantitet 50 mg
Synonym CK2 Inhibitor Casein kinase II Inhibitor
Löslighetsinformation DMSO : 50 mg/mL (104.87 mM; Need ultrasonic)
LEDER CN(C)C1=NC2=C(Br)C(Br)=C(Br)C(Br)=C2N1
Molekylvikt (g/mol) 476.79
Formel vikt 476.79
Procent renhet 98.03%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease