Learn More
Tocris Bioscience™ DMH2
Type I BMP receptor inhibitor
Brand:  Tocris Bioscience™ 5580/10
Additional Details : CAS Number : 1206711-14-9 Weight : 0.00970kg
 View more versions of this product
Product Code. 18716158
  • 2913.53 SEK / 10mg
Estimated Shipment: 31-07-2024
to see stock.
Description
Description
DMH2 is a type I BMP receptor inhibitor (Ki values are <1, 5.4 and 43 nM for ALK6, ALK3 and ALK2, respectively). Exhibits >30-fold selectivity over ALK4, ALK5, BMPR2, AMPK and VEGFR2. Also binds to TGFBR2 (Ki = 85 nM). Decreases growth and induces cell death of H1299 non-small cell lung carcinoma cells in vitro. Enhances liver regeneration in vivo.
Specifications
Specifications
4-[6-[4-[2-(4-Morpholinyl)ethoxy]phenyl]pyrazolo[1,5-a]pyrimidin-3-yl]quinoline
10 mg
C27H25N5O2
>98%
1206711-14-9
BMP and Other Activin Receptor Inhibitors
451.52