missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DMH2

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
DMH2 is a type I BMP receptor inhibitor (Ki values are <1, 5.4 and 43 nM for ALK6, ALK3 and ALK2, respectively). Exhibits >30-fold selectivity over ALK4, ALK5, BMPR2, AMPK and VEGFR2. Also binds to TGFBR2 (Ki = 85 nM). Decreases growth and induces cell death of H1299 non-small cell lung carcinoma cells in vitro. Enhances liver regeneration in vivo.
Greener Choice Claims
- Environmental benefits include:
- Ozone-Safe or Safe for the Ozone Layer

Specifications
Specifications
Chemical Name or Material | 4-[6-[4-[2-(4-Morpholinyl)ethoxy]phenyl]pyrazolo[1,5-a]pyrimidin-3-yl]quinoline |
CAS | 1206711-14-9 |
Quantity | 50 mg |
Target | BMP and Other Activin Receptor Inhibitors |
Molecular Formula | C27H25N5O2 |
Formula Weight | 451.52 |
Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tocris Bioscience™ DMH2 >
Spot an opportunity for improvement?Share a Content Correction